Assay Details
| Gene Symbol: Masp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008555.2 | NP_032581.2 | 5 - 6 | 793 | 101 | |
| GenBank mRNA | AB049755.1 | - | 5 - 6 | 806 | 101 | |
| AK031598.1 | - | 5 - 6 | 854 | 101 | ||
| AK043055.1 | - | 5 - 6 | 853 | 101 | ||
| BC131638.1 | - | 5 - 6 | 793 | 40109639 | 101 | |
| D16492.1 | - | 806 | 101 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Masp1
Genomic Map