Assay Details
| Gene Symbol: Eif2b5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172265.2 | NP_758469.1 | 4 - 5 | 772 | 91 | |
| GenBank mRNA | AK145801.1 | - | 4 - 5 | 774 | 91 | |
| AK160985.1 | - | 4 - 5 | 701 | 91 | ||
| BC027268.1 | - | 4 - 5 | 719 | 91 | ||
| BC038620.1 | - | 4 - 5 | 772 | 4211776 | 91 | |
| BC085255.1 | - | 4 - 5 | 683 | 5342410 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eif2b5
Genomic Map