Assay Details
Gene Symbol: Atp6v1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007508.5 | NP_031534.2 | 2 - 3 | 155 | 98 | |
GenBank mRNA | AK140873.1 | - | 2 - 3 | 131 | 98 | |
AK149833.1 | - | 2 - 3 | 170 | 98 | ||
AK150742.1 | - | 2 - 3 | 141 | 98 | ||
AK153403.1 | - | 2 - 3 | 156 | 98 | ||
AK154869.1 | - | 2 - 3 | 156 | 98 | ||
AK160792.1 | - | 2 - 3 | 170 | 98 | ||
AK166857.1 | - | 2 - 3 | 145 | 98 | ||
AK170721.1 | - | 2 - 3 | 156 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
