Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600005789.mm.1 |
NM_145972.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600005789.mm.1 |
NM_145972.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600005789.mm.1 |
TC1600000659.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600005791.mm.1 |
NM_145972.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600005791.mm.1 |
NM_145972.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600005791.mm.1 |
TC1600000659.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600005790.mm.1 |
NM_145972.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600005790.mm.1 |
NM_145972.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600005790.mm.1 |
TC1600000659.mm.1 |
MTA TC |