Assay Details
| Gene Symbol: Sidt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159419.1 | NP_001152891.1 | 15 - 16 | 1792 | 130 | |
| NM_198034.3 | NP_932151.2 | 15 - 16 | 1792 | 130 | ||
| GenBank mRNA | BC025888.1 | - | 5 - 6 | 442 | 5039343 | 130 |
| BC079584.1 | - | 15 - 16 | 1792 | 5704672 | 130 | |
More Information
Set Membership: |
|

Sidt1
Genomic Map