Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600006195.mm.1 |
NM_028680.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600006195.mm.1 |
NM_028680.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600006195.mm.1 |
TC1600000704.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600006193.mm.1 |
NM_028680.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600006193.mm.1 |
NM_028680.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600006193.mm.1 |
TC1600000704.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600006194.mm.1 |
NM_028680.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600006194.mm.1 |
NM_028680.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600006194.mm.1 |
TC1600000704.mm.1 |
MTA TC |