Assay Details
| Gene Symbol: Jam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023844.5 | NP_076333.3 | 2 - 3 | 499 | 82 | |
| GenBank mRNA | AF255911.1 | - | 2 - 3 | 222 | 82 | |
| AJ291757.1 | - | 2 - 3 | 435 | 82 | ||
| AK010616.1 | - | 2 - 3 | 341 | 82 | ||
| AK010826.1 | - | 2 - 3 | 343 | 82 | ||
| AK013914.1 | - | 2 - 3 | 340 | 82 | ||
| AK028757.1 | - | 2 - 3 | 190 | 82 | ||
| AK045095.1 | - | 2 - 3 | 237 | 82 | ||
| AK078128.1 | - | 2 - 3 | 225 | 82 | ||
| BC028778.1 | - | 2 - 3 | 499 | 1195543 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Jam2
Genomic Map