Assay Details
| Gene Symbol: Jam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023844.5 | NP_076333.3 | 4 - 5 | 764 | 66 | |
| GenBank mRNA | AF255911.1 | - | 4 - 5 | 487 | 66 | |
| AJ291757.1 | - | 4 - 5 | 700 | 66 | ||
| AK010616.1 | - | 4 - 5 | 606 | 66 | ||
| AK013914.1 | - | 4 - 5 | 605 | 66 | ||
| AK078128.1 | - | 4 - 5 | 490 | 66 | ||
| AK153636.1 | - | 4 - 5 | 66 | |||
| BC028778.1 | - | 4 - 5 | 764 | 1195543 | 66 | |
More Information
Set Membership: |
|

Jam2
Genomic Map