Assay Details
Gene Symbol: Crybg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174848.3 | NP_777273.2 | 14 - 15 | 3045 | 92 | |
GenBank mRNA | AK030282.1 | - | 5 - 6 | 489 | 92 | |
AK162506.1 | - | 2 - 3 | 880 | 92 | ||
BC043118.1 | - | 14 - 15 | 3045 | 6408859 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
