Assay Details
| Gene Symbol: Arhgap31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020260.2 | NP_064656.2 | 6 - 7 | 1040 | 113 | |
| GenBank mRNA | AF151363.1 | - | 6 - 7 | 681 | 113 | |
| AK138417.1 | - | 6 - 7 | 1280 | 113 | ||
| AK149975.1 | - | 6 - 7 | 876 | 113 | ||
| AK150109.1 | - | 6 - 7 | 876 | 113 | ||
| AK155729.1 | - | 6 - 7 | 860 | 113 | ||
| AK170685.1 | - | 6 - 7 | 860 | 113 | ||
| BC138870.1 | - | 6 - 7 | 845 | 8861892 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap31
Genomic Map