Assay Details
| Gene Symbol: Sod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011434.1 | NP_035564.1 | 2 - 3 | 281 | 89 | |
| GenBank mRNA | AK020624.1 | - | 2 - 3 | 187 | 89 | |
| AK077284.1 | - | 2 - 3 | 221 | 89 | ||
| BC002066.1 | - | 2 - 3 | 235 | 3496016 | 89 | |
| BC032986.1 | - | 3 - 4 | 89 | |||
| BC048874.1 | - | 2 - 3 | 281 | 5401293 | 89 | |
| BC086886.1 | - | 2 - 3 | 273 | 6760550 | 89 | |
| M35725.1 | - | 2 - 3 | 169 | 89 | ||
| X06683.1 | - | 2 - 3 | 169 | 89 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sod1
Genomic Map