Assay Details
| Gene Symbol: Psmd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134101.2 | NP_598862.1 | 2 - 3 | 256 | 86 | |
| NR_027485.1 | 2 - 3 | 348 | 86 | |||
| GenBank mRNA | AK146482.1 | - | 2 - 3 | 205 | 86 | |
| AK150374.1 | - | 2 - 3 | 228 | 86 | ||
| AK151588.1 | - | 2 - 3 | 228 | 86 | ||
| AK152007.1 | - | 2 - 3 | 228 | 86 | ||
| AK152970.1 | - | 2 - 3 | 225 | 86 | ||
| AK159098.1 | - | 2 - 3 | 208 | 86 | ||
| AK159636.1 | - | 2 - 3 | 205 | 86 | ||
| AK159756.1 | - | 2 - 3 | 228 | 86 | ||
| AK159815.1 | - | 2 - 3 | 224 | 86 | ||
| AK166816.1 | - | 2 - 3 | 208 | 86 | ||
| AK167326.1 | - | 2 - 3 | 208 | 86 | ||
| AK167774.1 | - | 2 - 3 | 205 | 86 | ||
| AK167786.1 | - | 2 - 3 | 206 | 86 | ||
| AK167909.1 | - | 2 - 3 | 216 | 86 | ||
| AK167995.1 | - | 2 - 3 | 224 | 86 | ||
| AK168131.1 | - | 2 - 3 | 215 | 86 | ||
| AK168398.1 | - | 2 - 3 | 208 | 86 | ||
| AK168543.1 | - | 2 - 3 | 205 | 86 | ||
| AK169867.1 | - | 2 - 3 | 208 | 86 | ||
| AK180842.1 | - | 2 - 3 | 84 | 86 | ||
| AK212329.1 | - | 2 - 3 | 84 | 86 | ||
| BC021508.1 | - | 2 - 3 | 225 | 5347139 | 86 | |
| BC023049.1 | - | 2 - 3 | 173 | 5356268 | 86 | |
| BC024058.1 | - | 2 - 3 | 198 | 86 | ||
| BC024830.1 | - | 2 - 3 | 196 | 5358424 | 86 | |
| BC027388.1 | - | 2 - 3 | 220 | 4952935 | 86 | |
| BC027767.1 | - | 2 - 3 | 224 | 5347281 | 86 | |
| BC027822.1 | - | 2 - 3 | 207 | 5321505 | 86 | |
| BC028851.1 | - | 2 - 3 | 196 | 3660169 | 86 | |
| BC031128.1 | - | 2 - 3 | 225 | 4982313 | 86 | |
| BC033325.1 | - | 2 - 3 | 285 | 86 | ||
| BY307723.1 | - | 2 - 3 | 257 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC023049.1 transcripts.
Set Membership: |
|

Psmd2
Genomic Map