Assay Details
Gene Symbol: Ntan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010946.3 | NP_035076.1 | 3 - 4 | 322 | 75 | |
GenBank mRNA | AK041112.1 | - | 3 - 4 | 279 | 75 | |
AK214245.1 | - | 1 - 2 | 70 | 75 | ||
BC030172.1 | - | 3 - 4 | 320 | 5037501 | 75 | |
BY168976.1 | - | 3 - 4 | 284 | 75 | ||
U57692.1 | - | 3 - 4 | 276 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
