Assay Details
Gene Symbol: Ntan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010946.3 | NP_035076.1 | 5 - 6 | 513 | 144 | |
GenBank mRNA | AK041112.1 | - | 5 - 6 | 470 | 144 | |
AK207258.1 | - | 1 - 2 | 61 | 144 | ||
AK214245.1 | - | 3 - 4 | 261 | 144 | ||
BC030172.1 | - | 5 - 6 | 511 | 5037501 | 144 | |
U57692.1 | - | 5 - 6 | 467 | 144 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
