Assay Details
| Gene Symbol: Pdxdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039533.2 | NP_001034622.1 | 17 - 18 | 1780 | 57 | |
| NM_001291017.1 | NP_001277946.1 | 17 - 18 | 1780 | 57 | ||
| NM_053181.3 | NP_444411.2 | 17 - 18 | 1780 | 57 | ||
| GenBank mRNA | AK004611.1 | - | 17 - 18 | 1657 | 57 | |
| AK030613.1 | - | 17 - 18 | 1611 | 57 | ||
| AK033107.1 | - | 17 - 18 | 1611 | 57 | ||
| AK054329.1 | - | 17 - 18 | 1763 | 57 | ||
| AK075829.1 | - | 10 - 11 | 903 | 57 | ||
| AK163696.1 | - | 17 - 18 | 1651 | 57 | ||
| AK165587.1 | - | 17 - 18 | 1615 | 57 | ||
| AK207349.1 | - | 2 - 3 | 283 | 57 | ||
| AK209827.1 | - | 2 - 3 | 283 | 57 | ||
| AK217692.1 | - | 2 - 3 | 283 | 57 | ||
| BC005541.1 | - | 17 - 18 | 1598 | 3590316 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pdxdc1
Genomic Map