Assay Details
Gene Symbol: Pdxdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039533.2 | NP_001034622.1 | 9 - 10 | 1028 | 66 | |
NM_001291017.1 | NP_001277946.1 | 9 - 10 | 1028 | 66 | ||
NM_053181.3 | NP_444411.2 | 9 - 10 | 1028 | 66 | ||
GenBank mRNA | AK004611.1 | - | 9 - 10 | 905 | 66 | |
AK030613.1 | - | 9 - 10 | 859 | 66 | ||
AK033107.1 | - | 9 - 10 | 859 | 66 | ||
AK054329.1 | - | 9 - 10 | 1011 | 66 | ||
AK075829.1 | - | 2 - 3 | 152 | 66 | ||
AK163696.1 | - | 9 - 10 | 899 | 66 | ||
AK165587.1 | - | 9 - 10 | 863 | 66 | ||
AK170661.1 | - | 9 - 10 | 1015 | 66 | ||
BC005541.1 | - | 9 - 10 | 846 | 3590316 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK075829.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
