Assay Details
| Gene Symbol: Marf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK035704.1 | - | 324 | 139 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK035704.1 transcripts.
Set Membership: |
|

Marf1
Genomic Map