Assay Details
| Gene Symbol: Rrn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039521.1 | NP_001034610.1 | 10 - 11 | 913 | 69 | |
| GenBank mRNA | AK080397.1 | - | 10 - 11 | 869 | 69 | |
| AK080680.1 | - | 894 | 69 | |||
| AK165676.1 | - | 10 - 11 | 902 | 69 | ||
| AK211678.1 | - | 1 - 2 | 76 | 69 | ||
| BC034110.1 | - | 6 - 7 | 428 | 69 | ||
| BC138774.1 | - | 10 - 11 | 878 | 8861796 | 69 | |
| BC138776.1 | - | 10 - 11 | 878 | 8861798 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rrn3
Genomic Map