Assay Details
| Gene Symbol: Rrn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039521.1 | NP_001034610.1 | 14 - 15 | 1498 | 77 | |
| GenBank mRNA | AK080397.1 | - | 14 - 15 | 1454 | 77 | |
| AK080680.1 | - | 1478 | 77 | |||
| AK165676.1 | - | 14 - 15 | 1486 | 77 | ||
| BC034110.1 | - | 10 - 11 | 1013 | 77 | ||
| BC055781.1 | - | 1 - 2 | 116 | 77 | ||
| BC138774.1 | - | 14 - 15 | 1463 | 8861796 | 77 | |
| BC138776.1 | - | 14 - 15 | 1463 | 8861798 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rrn3
Genomic Map