Assay Details
| Gene Symbol: Rrn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039521.1 | NP_001034610.1 | 3 - 4 | 302 | 139 | |
| GenBank mRNA | AK080397.1 | - | 3 - 4 | 257 | 139 | |
| AK080680.1 | - | 283 | 139 | |||
| AK165676.1 | - | 3 - 4 | 291 | 139 | ||
| BC138774.1 | - | 3 - 4 | 267 | 8861796 | 139 | |
| BC138776.1 | - | 3 - 4 | 267 | 8861798 | 139 | |
| CJ175904.1 | - | 3 - 4 | 305 | 139 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rrn3
Genomic Map