Assay Details
| Gene Symbol: Rrn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039521.1 | NP_001034610.1 | 8 - 9 | 713 | 102 | |
| GenBank mRNA | AK080397.1 | - | 8 - 9 | 668 | 102 | |
| AK080680.1 | - | 694 | 102 | |||
| AK165676.1 | - | 8 - 9 | 702 | 102 | ||
| BC034110.1 | - | 4 - 5 | 228 | 102 | ||
| BC138774.1 | - | 8 - 9 | 678 | 8861796 | 102 | |
| BC138776.1 | - | 8 - 9 | 678 | 8861798 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rrn3
Genomic Map