Assay Details
| Gene Symbol: Ncbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026554.4 | NP_080830.1 | 1 - 2 | 184 | 104 | |
| GenBank mRNA | AK008980.1 | - | 1 - 2 | 124 | 104 | |
| AK012659.1 | - | 1 - 2 | 104 | 104 | ||
| AK077841.1 | - | 1 - 2 | 92 | 104 | ||
| AK135597.1 | - | 1 - 2 | 98 | 104 | ||
| AK147027.1 | - | 1 - 2 | 116 | 104 | ||
| BC118926.2 | - | 1 - 2 | 92 | 40090166 | 104 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ncbp2
Genomic Map