Assay Details
| Gene Symbol: N6amt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159331.1 | NP_001152803.1 | 2 - 3 | 294 | 77 | |
| NM_026366.2 | NP_080642.1 | 2 - 3 | 294 | 77 | ||
| GenBank mRNA | AK012019.1 | - | 2 - 3 | 234 | 77 | |
| AK145617.1 | - | 2 - 3 | 246 | 77 | ||
| AK160298.1 | - | 2 - 3 | 256 | 77 | ||
| AY456393.1 | - | 2 - 3 | 294 | 77 | ||
| AY536887.1 | - | 2 - 3 | 226 | 77 | ||
| AY583759.1 | - | 2 - 3 | 338 | 77 | ||
| BC116393.1 | - | 2 - 3 | 264 | 40101877 | 77 | |
| BC116394.1 | - | 2 - 3 | 264 | 40101881 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

N6amt1
Genomic Map