Assay Details
| Gene Symbol: Ncam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113208.1 | NP_001106679.1 | 11 - 12 | 1696 | 103 | |
| NM_010954.4 | NP_035084.1 | 11 - 12 | 1696 | 103 | ||
| GenBank mRNA | AF001286.1 | - | 11 - 12 | 1622 | 103 | |
| AF001287.1 | - | 11 - 12 | 1622 | 103 | ||
| AF016619.1 | - | 11 - 12 | 1578 | 103 | ||
| BC119027.1 | - | 11 - 12 | 1554 | 8733776 | 103 | |
| BC119029.1 | - | 11 - 12 | 1554 | 8733778 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ncam2
Genomic Map