Assay Details
Gene Symbol: Gmip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198101.1 | NP_932769.1 | 16 - 17 | 1863 | 67 | |
GenBank mRNA | AK019874.2 | - | 5 - 6 | 613 | 67 | |
AK052221.1 | - | 16 - 17 | 1828 | 67 | ||
AK082337.1 | - | 16 - 17 | 1860 | 67 | ||
AK137177.1 | - | 14 - 15 | 1856 | 67 | ||
AK172438.1 | - | 16 - 17 | 1851 | 67 | ||
BC057037.1 | - | 16 - 17 | 1863 | 5702855 | 67 | |
BC060628.1 | - | 16 - 17 | 1838 | 6847400 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
