Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500024013.mm.1 |
TC0500002722.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500024013.mm.1 |
NM_001033310.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024013.mm.1 |
NM_001033310.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024013.mm.1 |
NM_001163456.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024013.mm.1 |
NM_001163456.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024013.mm.1 |
NR_028088.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024013.mm.1 |
NR_028088.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024012.mm.1 |
TC0500002722.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500024012.mm.1 |
NM_001033310.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024012.mm.1 |
NM_001033310.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024012.mm.1 |
NM_001163456.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024012.mm.1 |
NM_001163456.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024012.mm.1 |
NR_028088.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024012.mm.1 |
NR_028088.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024010.mm.1 |
TC0500002722.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500024010.mm.1 |
NM_001033310.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024010.mm.1 |
NM_001033310.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024010.mm.1 |
NM_001163456.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024010.mm.1 |
NM_001163456.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024010.mm.1 |
NR_028088.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024010.mm.1 |
NR_028088.1 |
Refseq |