Assay Details
Gene Symbol: Cox18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033310.3 | NP_001028482.2 | 4 - 5 | 764 | 81 | |
NM_001163456.1 | NP_001156928.1 | 4 - 5 | 767 | 81 | ||
NR_028088.1 | 4 - 5 | 768 | 81 | |||
GenBank mRNA | AK156053.1 | - | 4 - 5 | 733 | 81 | |
BC021612.1 | - | 2 - 3 | 315 | 81 | ||
BC038311.1 | - | 4 - 5 | 537 | 81 | ||
BC138972.1 | - | 4 - 5 | 742 | 8861994 | 81 | |
BC145479.1 | - | 4 - 5 | 745 | 9054042 | 81 | |
BC145486.1 | - | 4 - 5 | 746 | 81 | ||
BC145728.1 | - | 4 - 5 | 742 | 40131031 | 81 | |
DV661727.1 | - | 3 - 4 | 375 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23881742 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23881744 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23881741 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23881745 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23881743 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990134 | BE690958.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990134 | AK034738.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990134 | AK156053.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990134 | BC021612.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990134 | BC038311.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 835293 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 729249 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 437251 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 864882 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 4536 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P173244 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990134 | BE690958.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990134 | AK034738.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990134 | AK156053.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990134 | BC021612.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990134 | BC038311.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23881743 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23881744 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23881780 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23881735 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23881778 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23881770 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23881787 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23881746 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23881762 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23881736 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23881734 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23881732 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23881785 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23881741 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23881730 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23881753 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23881769 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23881748 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23881781 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23881756 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23881786 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23881759 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23881773 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23881764 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23881765 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23881767 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23881784 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23881776 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23881760 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23881772 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23881738 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23881750 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23881788 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23881754 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23881728 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05650757 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P173244 | - | - |
Whole Mouse Genome G4122A | A_52_P665768 | BE690958.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P665768 | AK034738.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P665768 | NM_001033310.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P665768 | NM_001163456.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P665768 | NR_028088.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P665767 | BE690958.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P665767 | AK034738.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P665767 | NM_001033310.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P665767 | NM_001163456.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P665767 | NR_028088.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138387 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P173244 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23881743 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23881745 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23881744 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23881742 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23881741 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881678 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881756 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881695 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881726 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881753 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881738 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881658 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881700 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881725 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881679 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881655 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881713 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881696 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881730 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881694 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881708 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881672 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881735 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881743 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881652 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881748 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881716 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881646 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881750 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881702 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881650 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881647 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881707 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881699 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881709 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881661 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881649 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881767 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881684 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881718 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881744 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881651 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881711 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881764 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881692 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881705 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881673 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881724 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881741 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881653 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881728 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881639 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881664 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881754 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881765 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881682 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881736 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881759 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881638 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881704 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881636 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881641 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881697 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881681 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23881720 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04209146 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23881718 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23881724 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23881730 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04430796 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23881702 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04755599 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23881720 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23881695 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23881750 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23881762 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04430805 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23881728 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23881734 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23881700 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23881726 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23881765 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23881754 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04710313 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04430812 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04419730 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04755597 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23881708 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23881764 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23881704 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23881699 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23881727 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23881753 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23881756 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23881716 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23881759 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23881698 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04879502 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04755595 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04152953 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23881735 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23881709 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23881767 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04430795 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23881692 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23881710 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23881732 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23881713 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23881736 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10531209 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10531211 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10531210 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10531213 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10531214 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10531212 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1415710_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10531208 | - | - |
Mouse Genome 430 2.0 Array | 1415710_at | - | - |
Mouse Genome 430 2.0 Array | 1415710_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 231430_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 231430_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0500023998.mm.1 | TC0500002722.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500023998.mm.1 | NM_001033310.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500023998.mm.1 | NM_001033310.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500023998.mm.1 | NM_001163456.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500023998.mm.1 | NM_001163456.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500023998.mm.1 | NR_028088.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500023998.mm.1 | NR_028088.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500024003.mm.1 | TC0500002722.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500024003.mm.1 | NM_001033310.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500024003.mm.1 | NM_001033310.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500024003.mm.1 | NM_001163456.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500024003.mm.1 | NM_001163456.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500024003.mm.1 | NR_028088.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500024003.mm.1 | NR_028088.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500023997.mm.1 | TC0500002722.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500023997.mm.1 | NM_001033310.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500023997.mm.1 | NM_001033310.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500023997.mm.1 | NM_001163456.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500023997.mm.1 | NM_001163456.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500023997.mm.1 | NR_028088.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500023997.mm.1 | NR_028088.1 | Refseq |
Mouse Expression Set 430 | 1415710_PM_at | - | - |
Mouse Expression Set 430 | 1415710_at | - | - |
Mouse Expression Set 430 | 1415710_at | - | - |
Mouse Expression Set 430 | 1415710_PM_at | - | - |
Murine Genome U74v2 Set | 162029_PM_r_at | - | - |
Murine Genome U74v2 Set | 92388_at | - | - |
Murine Genome U74v2 Set | 162029_r_at | - | - |
Murine Genome U74v2 Set | 92388_PM_at | - | - |
Murine Genome U74v2 Set | 170220_r_at | - | - |
Murine Genome U74v2 Set | 170220_PM_r_at | - | - |
Mouse Genome 430A 2.0 Array | 1415710_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1415710_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1415710_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1415710_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1415710_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1415710_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 912436 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_2633039 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3002011 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3002011 | - | - |
