Assay Details
| Gene Symbol: 4933404G15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045920.1 | 4 - 5 | 1008 | 60 | ||
| GenBank mRNA | AK016605.1 | - | 4 - 5 | 984 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

4933404G15Rik
Genomic Map