Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500024204.mm.1 |
TC0500002745.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500024204.mm.1 |
NM_001080794.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024204.mm.1 |
NM_001080795.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024204.mm.1 |
NM_001080796.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024204.mm.1 |
NM_001080797.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024204.mm.1 |
NM_011816.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024203.mm.1 |
TC0500002745.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500024203.mm.1 |
NM_001080794.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024203.mm.1 |
NM_001080795.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024203.mm.1 |
NM_001080796.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024203.mm.1 |
NM_001080797.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024203.mm.1 |
NM_011816.4 |
Refseq |