Assay Details
Gene Symbol: Pcsk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013628.2 | NP_038656.1 | 2 - 3 | 357 | 135 | |
GenBank mRNA | AK043742.1 | - | 2 - 3 | 429 | 135 | |
BC108982.1 | - | 2 - 3 | 335 | 40047652 | 135 | |
BC108983.1 | - | 2 - 3 | 335 | 40047655 | 135 | |
M58589.1 | - | 2 - 3 | 357 | 135 | ||
M69196.1 | - | 2 - 3 | 463 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
