Assay Details
Gene Symbol: Pcsk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013628.2 | NP_038656.1 | 5 - 6 | 698 | 69 | |
GenBank mRNA | AK043742.1 | - | 5 - 6 | 770 | 69 | |
BC108982.1 | - | 5 - 6 | 676 | 40047652 | 69 | |
BC108983.1 | - | 5 - 6 | 676 | 40047655 | 69 | |
M69196.1 | - | 5 - 6 | 804 | 69 | ||
X57088.1 | - | 5 - 6 | 810 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
