Assay Details
| Gene Symbol: Stra6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK004855.1 | - | 14 - 15 | 1396 | 65 | |
| AK045033.1 | - | 15 - 16 | 1824 | 65 | ||
| AK050152.1 | - | 14 - 15 | 1407 | 65 | ||
| AK050348.1 | - | 14 - 15 | 1403 | 65 | ||
| AK173193.1 | - | 3 - 4 | 5455 | 65 | ||
| BC111889.1 | - | 14 - 15 | 1333 | 40060643 | 65 | |
| BC112408.1 | - | 14 - 15 | 1333 | 40060646 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stra6l
Genomic Map