Assay Details
| Gene Symbol: Ncbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033201.3 | NP_001028373.2 | 7 - 8 | 1020 | 57 | |
| GenBank mRNA | AK133991.1 | - | 7 - 8 | 976 | 57 | |
| AK134334.1 | - | 7 - 8 | 1011 | 57 | ||
| AK166458.1 | - | 7 - 8 | 802 | 57 | ||
| AK169557.1 | - | 7 - 8 | 1021 | 57 | ||
| BC033473.1 | - | 6 - 7 | 636 | 57 | ||
| BC034658.1 | - | 4 - 5 | 462 | 57 | ||
| BC055777.1 | - | 4 - 5 | 446 | 57 | ||
| BC138898.1 | - | 7 - 8 | 891 | 8861920 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ncbp1
Genomic Map