Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400003964.mm.1 |
NM_001145924.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400003964.mm.1 |
NM_001145924.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400003964.mm.1 |
NM_001145925.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400003964.mm.1 |
NM_001145925.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400003964.mm.1 |
NM_028137.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400003964.mm.1 |
NM_028137.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400003964.mm.1 |
TC0400000532.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400003957.mm.1 |
NM_001145925.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400003957.mm.1 |
NM_001145925.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400003957.mm.1 |
TC0400000532.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400003965.mm.1 |
NM_001145924.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400003965.mm.1 |
NM_001145924.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400003965.mm.1 |
NM_001145925.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400003965.mm.1 |
NM_001145925.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400003965.mm.1 |
NM_028137.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400003965.mm.1 |
NM_028137.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400003965.mm.1 |
TC0400000532.mm.1 |
MTA TC |