Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400003974.mm.1 |
NM_021436.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400003974.mm.1 |
NM_021436.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400003974.mm.1 |
TC0400000535.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400003976.mm.1 |
NM_021436.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400003976.mm.1 |
NM_021436.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400003976.mm.1 |
TC0400000535.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400003977.mm.1 |
NM_021436.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400003977.mm.1 |
NM_021436.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400003977.mm.1 |
TC0400000535.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400003975.mm.1 |
NM_021436.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400003975.mm.1 |
NM_021436.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400003975.mm.1 |
TC0400000535.mm.1 |
MTA TC |