Assay Details
| Gene Symbol: Akap8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK004801.1 | - | 13 - 14 | 1661 | 105 | |
| AK089092.1 | - | 13 - 14 | 1668 | 105 | ||
| BC042602.1 | - | 13 - 14 | 2538 | 105 | ||
| BC054433.1 | - | 11 - 12 | 2235 | 105 | ||
| BC060170.1 | - | 13 - 14 | 2535 | 105 | ||
| BC060209.1 | - | 12 - 13 | 2151 | 105 | ||
| BC078448.1 | - | 12 - 13 | 2041 | 105 | ||
| BC125521.1 | - | 13 - 14 | 1651 | 40130036 | 105 | |
| BC137746.1 | - | 13 - 14 | 1651 | 8860765 | 105 | |
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31144951 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31144959 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31144955 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31144954 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31144956 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31144953 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31144958 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31144952 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31144957 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P106716 | BC060170.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P106716 | BC060209.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P106716 | AK077864.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P106716 | AK089092.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P106716 | BC042602.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 83072 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 291776 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 688038 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 79656 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 882094 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 329958 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 863837 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 689803 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 731810 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 151546 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 323135 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 161478 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 113865 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146069 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139154 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P106716 | BC060170.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P106716 | BC060209.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P106716 | AK077864.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P106716 | AK089092.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P106716 | BC042602.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P176549 | AB028920.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31144944 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31144945 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31144955 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31144959 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31144941 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31144947 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31144956 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31144954 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31144952 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31144953 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31144950 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31144958 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31144939 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31144943 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P139154 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P146069 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08760833 | - | - |
| Whole Mouse Genome G4122A | A_51_P176549 | AB028920.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P560817 | BC137746.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P560817 | BC054433.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P560817 | BC060170.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P560817 | BC060209.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P560817 | BC078448.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106716 | BC060170.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106716 | BC060209.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106716 | AK077864.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106716 | AK089092.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106716 | BC042602.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P139154 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P146069 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13125894 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31144958 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31144957 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31144956 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31144954 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31144951 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31144953 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31144952 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31144959 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31144955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31144955 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31144931 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31144901 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31144911 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31144934 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31144952 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31144955 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31144945 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31144941 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31144912 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31144921 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31144953 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31144943 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31144939 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31144906 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31144936 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31144938 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31144922 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31144933 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31144925 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31144947 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31144908 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31144958 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31144919 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31144920 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31144928 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31144917 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31144903 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13125887 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13125876 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31144926 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31144915 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31144959 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13261738 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13474835 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13092882 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31144918 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13125881 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13261750 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31144927 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31144930 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13286410 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10449849 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449846 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449845 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449848 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449841 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449850 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449840 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449844 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449847 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449851 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449842 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449853 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449843 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449852 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450983_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1433669_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10449839 | - | - |
| Mouse Genome 430 2.0 Array | 1433669_at | - | - |
| Mouse Genome 430 2.0 Array | 1450983_at | - | - |
| Mouse Genome 430 2.0 Array | 1450983_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1433669_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56399_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56399_PM_at | - | - |
| Mouse Expression Set 430 | 1433669_PM_at | - | - |
| Mouse Expression Set 430 | 1433669_at | - | - |
| Mouse Expression Set 430 | 1450983_at | - | - |
| Mouse Expression Set 430 | 1450983_PM_at | - | - |
| Murine Genome U74v2 Set | 95001_at | - | - |
| Murine Genome U74v2 Set | 94390_PM_at | - | - |
| Murine Genome U74v2 Set | 95001_PM_at | - | - |
| Murine Genome U74v2 Set | 161089_r_at | - | - |
| Murine Genome U74v2 Set | 161088_PM_r_at | - | - |
| Murine Genome U74v2 Set | 161596_PM_f_at | - | - |
| Murine Genome U74v2 Set | 94390_at | - | - |
| Murine Genome U74v2 Set | 161089_PM_r_at | - | - |
| Murine Genome U74v2 Set | 161596_f_at | - | - |
| Murine Genome U74v2 Set | 161088_r_at | - | - |
| Murine Genome U74v2 Set | 170044_s_at | - | - |
| Murine Genome U74v2 Set | 170044_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450983_at | - | - |
| Mouse Genome 430A 2.0 Array | 1433669_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450983_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1433669_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450983_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1433669_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450983_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1433669_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 101570059 | - | - |
| MouseWG-6 v2 Expression BeadChip | 4210026 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4210026 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 101570059 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004210026 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1238872 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1238872 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1217938 | - | - |
| mouse-6 v1.1 (Illumina) | scl056399.1_113-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101570059 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4210026 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl056399.1_113-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4210026 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101570059 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2920008 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2920008 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1217938 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl056399.1_113-S | - | - |

Akap8
Genomic Map