Assay Details
| Gene Symbol: Cyp4f17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101445.1 | NP_001094915.1 | 1 - 2 | 120 | 112 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001101445.1 transcripts.
Set Membership: |
|

Cyp4f17
Genomic Map