Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700016778.mm.1 |
NM_001271353.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016778.mm.1 |
NM_144855.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016778.mm.1 |
NM_178224.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016778.mm.1 |
TC1700001822.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700016771.mm.1 |
NM_001271353.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016771.mm.1 |
NM_144855.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016771.mm.1 |
NM_178224.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016771.mm.1 |
TC1700001822.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700016775.mm.1 |
NM_001271353.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016775.mm.1 |
NM_144855.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016775.mm.1 |
NM_178224.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016775.mm.1 |
TC1700001822.mm.1 |
MTA TC |