Assay Details
Gene Symbol: Sik1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010831.2 | NP_034961.2 | 12 - 13 | 1883 | 82 | |
GenBank mRNA | AK087322.1 | - | 5 - 6 | 800 | 82 | |
AK141817.1 | - | 12 - 13 | 1883 | 82 | ||
AK172427.1 | - | 11 - 12 | 2323 | 82 | ||
AK220203.1 | - | 10 - 11 | 1496 | 82 | ||
U11494.2 | - | 12 - 13 | 1875 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
