Assay Details
Gene Symbol: Sik1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010831.2 | NP_034961.2 | 7 - 8 | 891 | 59 | |
GenBank mRNA | AK141817.1 | - | 7 - 8 | 891 | 59 | |
AK172427.1 | - | 7 - 8 | 894 | 59 | ||
AK220203.1 | - | 5 - 6 | 504 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
