Assay Details
| Gene Symbol: Akap8l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017476.2 | NP_059504.2 | 6 - 7 | 999 | 89 | |
| GenBank mRNA | AB028921.1 | - | 6 - 7 | 992 | 89 | |
| AK011325.1 | - | 6 - 7 | 1004 | 89 | ||
| BC046801.1 | - | 6 - 7 | 988 | 6403966 | 89 | |
| BC051015.1 | - | 6 - 7 | 999 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Akap8l
Genomic Map