Assay Details
| Gene Symbol: Akap8l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017476.2 | NP_059504.2 | 9 - 10 | 1241 | 60 | |
| GenBank mRNA | AB028921.1 | - | 9 - 10 | 1237 | 60 | |
| AK011325.1 | - | 9 - 10 | 1246 | 60 | ||
| AK166526.1 | - | 10 - 11 | 1157 | 60 | ||
| BC046801.1 | - | 9 - 10 | 1230 | 6403966 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Akap8l
Genomic Map