Assay Details
Gene Symbol: Fgd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK030390.1 | - | 1 - 2 | 266 | 99 | |
BB617683.1 | - | 1 - 2 | 266 | 99 | ||
BC129902.1 | - | 1 - 2 | 203 | 40111637 | 99 | |
BC129903.1 | - | 1 - 2 | 204 | 40111639 | 99 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC129902.1, BC129903.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -



MicroArray Probes:
