| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10513000 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10512999 |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0400021680.mm.1 |
TC0400002668.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400021680.mm.1 |
NR_033139.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400021680.mm.1 |
NR_033139.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400021677.mm.1 |
TC0400002668.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400021677.mm.1 |
NR_033139.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400021677.mm.1 |
NR_033139.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400021677.mm.1 |
NR_033140.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400021677.mm.1 |
NR_033140.1 |
Refseq |