Assay Details
Gene Symbol: Krt20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023256.1 | NP_075745.1 | 4 - 5 | 869 | 67 | |
GenBank mRNA | AF473907.1 | - | 4 - 5 | 846 | 67 | |
AK018567.1 | - | 4 - 5 | 869 | 67 | ||
BC057172.1 | - | 4 - 5 | 852 | 4989154 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
