Assay Details
Gene Symbol: Krt20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023256.1 | NP_075745.1 | 7 - 8 | 1253 | 66 | |
GenBank mRNA | AF473907.1 | - | 7 - 8 | 1230 | 66 | |
AK018567.1 | - | 7 - 8 | 1253 | 66 | ||
BC038900.1 | - | 3 - 4 | 340 | 66 | ||
BC057172.1 | - | 7 - 8 | 1236 | 4989154 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P445360 | AK018567.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P445360 | BC038900.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P445360 | BC057172.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P445360 | NM_023256.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P445360 | AK008079.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P445360 | AK018567.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P445360 | BC038900.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P445360 | BC057172.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P445360 | NM_023256.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P445360 | AK008079.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 218270 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 587272 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 6588 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 703652 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105748 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P445360 | AK018567.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P445360 | BC038900.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P445360 | BC057172.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P445360 | NM_023256.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P445360 | AK008079.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P445360 | AK018567.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P445360 | BC038900.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P445360 | BC057172.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P445360 | NM_023256.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P445360 | AK008079.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062367 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062380 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062386 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062382 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062356 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062365 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062342 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062361 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062388 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062374 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062363 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062369 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062359 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062347 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062377 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062344 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062348 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062371 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062353 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062351 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062372 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062341 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062385 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062357 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062376 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062350 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28062345 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07461168 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P105748 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07461186 | - | - |
Whole Mouse Genome G4122A | A_52_P445360 | AK018567.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P445360 | BC038900.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P445360 | BC057172.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P445360 | NM_023256.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P445360 | AK008079.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P445360 | AK018567.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P445360 | BC038900.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P445360 | BC057172.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P445360 | NM_023256.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P445360 | AK008079.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127677 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P105748 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09193800 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09107656 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09193794 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09193787 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09193797 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09193796 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062349 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062409 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062306 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062288 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062380 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062263 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062304 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062289 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062277 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062320 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062224 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062348 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062377 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062359 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062400 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062260 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062242 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062226 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062389 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062319 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062266 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062376 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062294 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062256 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062296 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062299 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062316 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062357 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062344 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062351 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062369 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062227 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062246 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062303 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062411 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062326 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062223 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062237 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062317 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062312 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062276 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062329 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062308 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062363 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062238 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062410 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062408 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062297 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062245 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062232 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062250 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062367 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062273 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062397 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062339 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062286 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062218 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062275 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062293 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062353 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062236 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062391 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062361 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062395 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062243 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062343 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062325 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062310 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062252 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062365 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062248 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062280 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062331 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062382 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062281 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062271 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062269 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062334 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062372 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062324 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062229 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062282 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062341 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062322 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062332 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062254 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062356 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062386 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28062371 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062331 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062389 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062294 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09359172 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09577378 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09162626 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062343 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062299 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062337 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062349 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062382 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09107656 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062351 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09671715 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062380 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062335 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09671686 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062371 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09682386 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09489796 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062324 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062318 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062327 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09016689 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09671714 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062319 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062376 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062312 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062353 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062393 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062397 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09162623 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09142301 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062369 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062386 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062404 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09193796 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062359 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062403 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062411 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062364 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062326 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062408 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062308 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062367 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09237682 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062377 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062329 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062316 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062293 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062388 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062348 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09359175 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09162624 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062400 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062357 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062336 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062399 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062320 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062332 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09016691 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09577372 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062372 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09706666 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062394 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062387 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062297 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09577371 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062322 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062361 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062345 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28062409 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10390853 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390856 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390851 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390857 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390855 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390858 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390852 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390859 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390854 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1426284_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10390850 | - | - |
Mouse Genome 430 2.0 Array | 1426284_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1426284_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66809_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66809_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100034217.mm.1 | NM_023256.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100034217.mm.1 | TC1100003708.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100034216.mm.1 | NM_023256.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100034216.mm.1 | TC1100003708.mm.1 | MTA TC |
Mouse Expression Set 430 | 1426284_at | - | - |
Mouse Expression Set 430 | 1426284_PM_at | - | - |
Murine Genome U74v2 Set | 95621_PM_at | - | - |
Murine Genome U74v2 Set | 95621_at | - | - |
Mouse Genome 430A 2.0 Array | 1426284_at | - | - |
Mouse Genome 430A 2.0 Array | 1426284_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1426284_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1426284_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 796270 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 4060193 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 4060193 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004060193 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2735093 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2735093 | - | - |
mouse-6 v1.1 (Illumina) | scl066809.1_67-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4060193 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl066809.1_67-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4060193 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2907285 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2907285 | - | - |
