Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400004594.mm.1 |
NM_001099323.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004594.mm.1 |
NM_001099323.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004594.mm.1 |
TC0400000650.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400004593.mm.1 |
NM_001099323.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004593.mm.1 |
NM_001099323.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004593.mm.1 |
TC0400000650.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400004592.mm.1 |
NM_001099323.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004592.mm.1 |
NM_001099323.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004592.mm.1 |
TC0400000650.mm.1 |
MTA TC |