Assay Details
| Gene Symbol: Pfn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029303.2 | NP_083579.1 | 1 | 494 | 104 | |
| GenBank mRNA | AK005930.1 | - | 1 | 486 | 104 | |
| BC048629.1 | - | 1 | 494 | 6744051 | 104 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29054895 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29054896 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29054897 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29054893 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P444517 | NM_029303.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P444517 | AK005930.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P444517 | BC048629.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P444517 | NM_029303.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P444517 | AK005930.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P444517 | BC048629.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164191 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P444517 | NM_029303.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P444517 | AK005930.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P444517 | BC048629.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P444517 | NM_029303.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P444517 | AK005930.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P444517 | BC048629.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P444517 | NM_029303.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P444517 | AK005930.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P444517 | BC048629.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P444517 | NM_029303.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P444517 | AK005930.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P444517 | BC048629.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054900 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054898 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054892 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054893 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054896 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054901 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054902 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054890 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P164191 | - | - |
| Whole Mouse Genome G4122A | A_52_P444517 | NM_029303.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P444517 | AK005930.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P444517 | BC048629.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P444517 | NM_029303.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P444517 | AK005930.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P444517 | BC048629.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134730 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P164191 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10976529 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10976533 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29054895 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29054896 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29054893 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29054897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054930 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10457642 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29054900 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10976533 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29054901 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11099425 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29054893 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10976529 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10409448 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409447 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10405463 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409446 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453962_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10409445 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10405462 | - | - |
| Mouse Genome 430 2.0 Array | 1427627_at | - | - |
| Mouse Genome 430 2.0 Array | 1427627_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1453962_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1453962_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75477_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75477_PM_at | - | - |
| Mouse Expression Set 430 | 1453962_PM_at | - | - |
| Mouse Expression Set 430 | 1453962_at | - | - |
| Mouse Genome 430A 2.0 Array | 1453962_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1453962_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1453962_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1453962_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 412304 | - | - |
| ABI Mouse Genome Survey Microarray | 422919 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4760133 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4760133 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004760133 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2610314 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2610314 | - | - |
| mouse-6 v1.1 (Illumina) | scl0075477.1_320-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4760133 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0075477.1_320-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4760133 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2610314 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2610314 | - | - |

Pfn3
Genomic Map