Assay Details
| Gene Symbol: Prss38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045521.1 | NP_001038986.1 | 3 - 4 | 595 | 83 | |
| GenBank mRNA | AK145810.1 | - | 3 - 4 | 600 | 83 | |
| BC147400.1 | - | 3 - 4 | 591 | 9056455 | 83 | |
| BC147401.1 | - | 3 - 4 | 591 | 9056447 | 83 | |
More Information
Set Membership: |
|

Prss38
Genomic Map