Assay Details
| Gene Symbol: Cdkl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276315.1 | NP_001263244.1 | 1 - 2 | 502 | 89 | |
| GenBank mRNA | AB029067.1 | - | 1 - 2 | 490 | 89 | |
| AK030598.1 | - | 1 - 2 | 479 | 89 | ||
| AK139820.1 | - | 1 - 2 | 444 | 89 | ||
| AK145688.1 | - | 1 - 2 | 385 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB029067.1, AK030598.1, AK145688.1, NM_001276315.1 transcripts.
Set Membership: |
|

Cdkl2
Genomic Map